Assay Details
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716782.3 | XP_002716828.1 | 2 - 3 | 459 | 87 | |
XM_008267170.2 | XP_008265392.1 | 3 - 4 | 544 | 87 | ||
XM_008267171.2 | XP_008265393.1 | 4 - 5 | 592 | 87 | ||
XM_008267172.2 | XP_008265394.1 | 3 - 4 | 485 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
