Assay Details
Gene Symbol: CCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723866.3 | XP_002723912.2 | 1663 | 111 | ||
XM_008252708.2 | XP_008250930.1 | 1839 | 111 | |||
XM_017339990.1 | XP_017195479.1 | 2200 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
