Assay Details
Gene Symbol: ADGRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008263020.2 | XP_008261242.1 | 4 - 5 | 465 | 105 | |
XM_017345168.1 | XP_017200657.1 | 4 - 5 | 465 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
