Assay Details
Gene Symbol: ADGRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008263020.2 | XP_008261242.1 | 7 - 8 | 2114 | 86 | |
XM_008263021.2 | XP_008261243.1 | 6 - 7 | 1880 | 86 | ||
XM_017345168.1 | XP_017200657.1 | 7 - 8 | 2114 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
