Assay Details
Gene Symbol: MAPK1IP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008269683.2 | XP_008267905.1 | 2 - 3 | 174 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008269683.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
