Assay Details
Gene Symbol: PPM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716344.3 | XP_002716390.1 | 2 - 3 | 670 | 99 | |
XM_017346885.1 | XP_017202374.1 | 2 - 3 | 226 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
