Assay Details
Gene Symbol: RUBCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266732.2 | XP_008264954.1 | 5 - 6 | 764 | 54 | |
XM_008266733.2 | XP_008264955.1 | 5 - 6 | 764 | 54 | ||
XM_008266734.1 | XP_008264956.1 | 5 - 6 | 986 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
