Assay Details
Gene Symbol: LOC100343709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710450.3 | XP_002710496.1 | 1 - 2 | 134 | 84 | |
XM_008255595.2 | XP_008253817.1 | 1 - 2 | 134 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002710450.3, XM_008255595.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
