Assay Details
Gene Symbol: LOC100343709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710450.3 | XP_002710496.1 | 8 - 9 | 753 | 91 | |
XM_008255595.2 | XP_008253817.1 | 8 - 9 | 753 | 91 | ||
XM_008255596.2 | XP_008253818.1 | 8 - 9 | 707 | 91 | ||
XM_008255598.2 | XP_008253820.1 | 8 - 9 | 738 | 91 | ||
XM_017341361.1 | XP_017196850.1 | 8 - 9 | 645 | 91 | ||
XM_017341362.1 | XP_017196851.1 | 8 - 9 | 542 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
