Assay Details
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715202.3 | XP_002715248.1 | 2 - 3 | 411 | 67 | |
XM_008265525.2 | XP_008263747.1 | 3 - 4 | 433 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
