Assay Details
Gene Symbol: LRIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264700.2 | XP_008262922.1 | 2 - 3 | 2177 | 74 | |
XM_008264701.2 | XP_008262923.1 | 3 - 4 | 1703 | 74 | ||
XM_008264706.2 | XP_008262928.1 | 2 - 3 | 2171 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
