Assay Details
Gene Symbol: C17H14orf93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718074.3 | XP_002718120.1 | 3 - 4 | 1431 | 90 | |
XM_008269352.2 | XP_008267574.1 | 3 - 4 | 1440 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
