Assay Details
Gene Symbol: RASSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266370.2 | XP_008264592.1 | 1 | 219 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008266370.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
