Assay Details
Gene Symbol: FCSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008257599.2 | XP_008255821.1 | 4 - 5 | 642 | 59 | |
XM_008257600.2 | XP_008255822.1 | 4 - 5 | 743 | 59 | ||
XM_008257602.2 | XP_008255824.1 | 4 - 5 | 642 | 59 | ||
XM_008257603.1 | XP_008255825.1 | 2 - 3 | 136 | 59 | ||
XM_008257604.2 | XP_008255826.1 | 4 - 5 | 644 | 59 | ||
XM_008257605.2 | XP_008255827.1 | 4 - 5 | 643 | 59 | ||
XM_017342356.1 | XP_017197845.1 | 4 - 5 | 644 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008257603.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
