Assay Details
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274143.2 | XP_008272365.1 | 870 | 91 | ||
XM_008274144.2 | XP_008272366.1 | 926 | 91 | |||
XM_017350463.1 | XP_017205952.1 | 876 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
