Assay Details
Gene Symbol: MYL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710781.3 | XP_002710827.1 | 3 - 4 | 488 | 103 | |
XM_017341640.1 | XP_017197129.1 | 3 - 4 | 402 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
