Assay Details
Gene Symbol: POGLUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713017.3 | XP_002713063.1 | 4 - 5 | 883 | 127 | |
XM_017343585.1 | XP_017199074.1 | 4 - 5 | 675 | 127 | ||
XM_017343586.1 | XP_017199075.1 | 3 - 4 | 327 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() |
