Assay Details
Gene Symbol: NIBAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017350074.1 | XP_017205563.1 | 134 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017350074.1 transcripts.
Set Membership: |
![]() ![]() |
