Assay Details
Gene Symbol: PPP4R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719598.3 | XP_002719644.1 | 6 - 7 | 2150 | 68 | |
XM_017349436.1 | XP_017204925.1 | 6 - 7 | 2150 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
