Assay Details
Gene Symbol: MRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716695.3 | XP_002716741.2 | 1 | 179 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002716695.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
