Assay Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008250551.2 | XP_008248773.1 | 905 | 78 | ||
XM_008250552.2 | XP_008248774.1 | 227 | 78 | |||
XM_017338674.1 | XP_017194163.1 | 307 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
