Assay Details
Gene Symbol: IL3RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008248460.2 | XP_008246682.1 | 5 - 6 | 609 | 62 | |
XM_008248462.2 | XP_008246684.1 | 4 - 5 | 488 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
