Assay Details
Gene Symbol: DOP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274856.2 | XP_008273078.1 | 3695 | 80 | ||
XM_008274857.2 | XP_008273079.1 | 3695 | 80 | |||
XM_017350777.1 | XP_017206266.1 | 3697 | 80 |

More Information
Set Membership: |
![]() ![]() |
