Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713858.3 | XP_002713904.1 | 11 - 12 | 1544 | 92 | |
XM_017344333.1 | XP_017199822.1 | 10 - 11 | 1310 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
