Assay Details
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717559.3 | XP_002717605.1 | 1 - 2 | 286 | 92 | |
XM_017347836.1 | XP_017203325.1 | 1 - 2 | 286 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
