Assay Details
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008250128.2 | XP_008248350.1 | 491 | 94 | ||
XM_008250129.2 | XP_008248351.1 | 400 | 94 | |||
XM_008250130.2 | XP_008248352.2 | 248 | 94 | |||
XM_017338485.1 | XP_017193974.1 | 248 | 94 | |||
XM_017338486.1 | XP_017193975.1 | 248 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
