Assay Details
Gene Symbol: LOC100341774 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008257389.2 | XP_008255611.1 | 2 - 3 | 336 | 116 | |
XM_017342269.1 | XP_017197758.1 | 2 - 3 | 258 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
