Assay Details
Gene Symbol: TANK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008258660.2 | XP_008256882.1 | 6 - 7 | 693 | 90 | |
XM_008258661.2 | XP_008256883.1 | 6 - 7 | 684 | 90 | ||
XM_008258662.2 | XP_008256884.1 | 6 - 7 | 690 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
