Assay Details
Gene Symbol: TRIM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718818.3 | XP_002718864.1 | 5 - 6 | 1295 | 59 | |
XM_017348922.1 | XP_017204411.1 | 5 - 6 | 1373 | 59 | ||
XM_017348923.1 | XP_017204412.1 | 5 - 6 | 1376 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
