Assay Details
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008268892.2 | XP_008267114.1 | 7 - 8 | 903 | 67 | |
XM_008268897.2 | XP_008267119.1 | 7 - 8 | 785 | 67 | ||
XM_008268910.2 | XP_008267132.1 | 7 - 8 | 731 | 67 | ||
XM_017345590.1 | XP_017201079.1 | 7 - 8 | 834 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
