Assay Details
Gene Symbol: LOC100342021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008253649.2 | XP_008251871.1 | 1 - 2 | 194 | 115 | |
XM_008253656.2 | XP_008251878.1 | 1 - 2 | 205 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008253649.2, XM_008253656.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
