Assay Details
Gene Symbol: LOC100342021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008253649.2 | XP_008251871.1 | 2 - 3 | 640 | 73 | |
XM_008253656.2 | XP_008251878.1 | 2 - 3 | 730 | 73 | ||
XM_017339820.1 | XP_017195309.1 | 2 - 3 | 546 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
