Assay Details
Gene Symbol: LOC100342021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008253649.2 | XP_008251871.1 | 6 - 7 | 1073 | 72 | |
XM_008253656.2 | XP_008251878.1 | 6 - 7 | 1163 | 72 | ||
XM_017339820.1 | XP_017195309.1 | 6 - 7 | 979 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
