Assay Details
Gene Symbol: GPR171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266339.2 | XP_008264561.1 | 1 - 2 | 139 | 79 | |
XM_017346846.1 | XP_017202335.1 | 1 - 2 | 144 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008266339.2, XM_017346846.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
