Assay Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008263524.1 | XP_008261746.1 | 6 - 7 | 1022 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
