Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008268182.2 | XP_008266404.1 | 14 - 15 | 1990 | 138 | |
XM_008268183.2 | XP_008266405.1 | 14 - 15 | 1990 | 138 | ||
XM_008268184.2 | XP_008266406.1 | 13 - 14 | 1837 | 138 | ||
XM_008268188.2 | XP_008266410.1 | 15 - 16 | 1892 | 138 | ||
XM_008268191.2 | XP_008266413.1 | 15 - 16 | 1879 | 138 | ||
XM_008268192.2 | XP_008266414.1 | 14 - 15 | 1814 | 138 | ||
XM_008268193.2 | XP_008266415.1 | 12 - 13 | 1562 | 138 | ||
XM_017347704.1 | XP_017203193.1 | 15 - 16 | 1905 | 138 | ||
XM_017347705.1 | XP_017203194.1 | 15 - 16 | 1850 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
