Assay Details
Gene Symbol: MINDY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718209.3 | XP_002718255.1 | 3 - 4 | 1037 | 87 | |
XM_017347995.1 | XP_017203484.1 | 3 - 4 | 1037 | 87 | ||
XM_017347996.1 | XP_017203485.1 | 3 - 4 | 1037 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
