Assay Details
Gene Symbol: MINDY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718209.3 | XP_002718255.1 | 6 - 7 | 1442 | 114 | |
XM_017347995.1 | XP_017203484.1 | 6 - 7 | 1442 | 114 | ||
XM_017347996.1 | XP_017203485.1 | 6 - 7 | 1442 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
