Assay Details
Gene Symbol: VSIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720060.3 | XP_002720106.1 | 5 - 6 | 962 | 97 | |
XM_008272723.2 | XP_008270945.1 | 5 - 6 | 962 | 97 | ||
XM_008272724.2 | XP_008270946.1 | 5 - 6 | 962 | 97 | ||
XM_017349982.1 | XP_017205471.1 | 5 - 6 | 962 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
