Assay Details
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002721571.3 | XP_002721617.1 | 1416 | 62 | ||
XM_008248395.2 | XP_008246617.1 | 1549 | 62 | |||
XM_008248396.2 | XP_008246618.1 | 1416 | 62 | |||
XM_008248397.2 | XP_008246619.1 | 1439 | 62 | |||
XM_008248398.2 | XP_008246620.1 | 1549 | 62 | |||
XM_008248399.2 | XP_008246621.1 | 1444 | 62 | |||
XM_008248400.2 | XP_008246622.1 | 1246 | 62 | |||
XM_008248401.2 | XP_008246623.1 | 1113 | 62 | |||
XM_017337625.1 | XP_017193114.1 | 1500 | 62 | |||
XM_017337626.1 | XP_017193115.1 | 1450 | 62 | |||
XM_017337627.1 | XP_017193116.1 | 1147 | 62 | |||
XM_017337628.1 | XP_017193117.1 | 1141 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
