Assay Details
Gene Symbol: PDE4DIPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264567.2 | XP_008262789.2 | 28 - 29 | 4240 | 63 | |
XM_008264576.2 | XP_008262798.2 | 28 - 29 | 4240 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
