Assay Details
Gene Symbol: PDE4DIPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264566.2 | XP_008262788.2 | 31 - 32 | 5120 | 78 | |
XM_008264567.2 | XP_008262789.2 | 31 - 32 | 5116 | 78 | ||
XM_008264569.2 | XP_008262791.2 | 31 - 32 | 5120 | 78 | ||
XM_008264570.2 | XP_008262792.2 | 31 - 32 | 5120 | 78 | ||
XM_008264573.2 | XP_008262795.1 | 31 - 32 | 5174 | 78 | ||
XM_008264574.2 | XP_008262796.1 | 24 - 25 | 5356 | 78 | ||
XM_008264575.2 | XP_008262797.1 | 24 - 25 | 4103 | 78 | ||
XM_008264576.2 | XP_008262798.2 | 31 - 32 | 5116 | 78 | ||
XM_017345940.1 | XP_017201429.1 | 31 - 32 | 5121 | 78 | ||
XM_017345941.1 | XP_017201430.1 | 28 - 29 | 4880 | 78 | ||
XM_017345942.1 | XP_017201431.1 | 27 - 28 | 4518 | 78 | ||
XM_017345943.1 | XP_017201432.1 | 28 - 29 | 4880 | 78 | ||
XM_017345944.1 | XP_017201433.1 | 28 - 29 | 4880 | 78 | ||
XM_017345945.1 | XP_017201434.1 | 31 - 32 | 5121 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
