Assay Details
Gene Symbol: PDE4DIPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264566.2 | XP_008262788.2 | 3 - 4 | 505 | 80 | |
XM_008264567.2 | XP_008262789.2 | 3 - 4 | 504 | 80 | ||
XM_008264569.2 | XP_008262791.2 | 3 - 4 | 505 | 80 | ||
XM_008264570.2 | XP_008262792.2 | 3 - 4 | 505 | 80 | ||
XM_008264573.2 | XP_008262795.1 | 3 - 4 | 559 | 80 | ||
XM_008264576.2 | XP_008262798.2 | 3 - 4 | 504 | 80 | ||
XM_017345940.1 | XP_017201429.1 | 3 - 4 | 506 | 80 | ||
XM_017345945.1 | XP_017201434.1 | 3 - 4 | 506 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
