Assay Details
Gene Symbol: PKIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710648.2 | XP_002710694.2 | 2 - 3 | 279 | 83 | |
XM_008255683.2 | XP_008253905.1 | 3 - 4 | 442 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
