Assay Details
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719315.3 | XP_002719361.1 | 1 | 63 | 85 | |
XM_008271350.2 | XP_008269572.1 | 1 | 63 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002719315.3, XM_008271350.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
