Assay Details
Gene Symbol: YWHAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082168.2 | NP_001075637.2 | 1 | 330 | 73 | |
GenBank mRNA | EB377152.1 | - | 2 | 112 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
