Assay Details
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261512.2 | NP_001248441.1 | 326 | 111 | ||
XM_015151139.2 | XP_015006625.1 | 488 | 111 | |||
XM_015151140.2 | XP_015006626.1 | 468 | 111 | |||
XM_015151141.2 | XP_015006627.1 | 488 | 111 | |||
XM_028828910.1 | XP_028684743.1 | 468 | 111 | |||
GenBank mRNA | JU334438.1 | - | 346 | 111 | ||
JU475527.1 | - | 430 | 111 | |||
JU475528.1 | - | 301 | 111 | |||
JV047013.1 | - | 330 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
