Assay Details
Gene Symbol: GAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128499.1 | XP_014983985.1 | 122 | 79 | ||
XM_015128500.1 | XP_014983986.1 | 122 | 79 | |||
XM_015128501.1 | XP_014983987.1 | 122 | 79 | |||
XM_028841849.1 | XP_028697682.1 | 122 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
