Assay Details
Gene Symbol: TANK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257543.1 | NP_001244472.1 | 155 | 89 | ||
XM_015110164.2 | XP_014965650.1 | 683 | 89 | |||
XM_015110165.2 | XP_014965651.1 | 301 | 89 | |||
XM_015110166.2 | XP_014965652.1 | 344 | 89 | |||
XM_015110169.2 | XP_014965655.1 | 169 | 89 | |||
XM_028830463.1 | XP_028686296.1 | 694 | 89 | |||
XM_028830464.1 | XP_028686297.1 | 570 | 89 | |||
GenBank mRNA | JU322903.1 | - | 155 | 89 | ||
JU472929.1 | - | 265 | 89 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
