Assay Details
Gene Symbol: GABARAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194264.2 | NP_001181193.1 | 373 | 70 | ||
GenBank mRNA | JU472337.1 | - | 320 | 70 | ||
JV045121.1 | - | 368 | 70 | |||
JV453052.1 | - | 381 | 70 | |||
JV635645.1 | - | 358 | 70 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
